Learn More
Cayman Chemical ANTIBODY OF-02 1mg
SDP
Supplier:  Cayman Chemical 376521 MG
Encompass_Preferred
An ionizable cationic amino lipid; has been used in combination with other lipids in the formation of LNPs for the delivery of mRNA; LNPs containing OF-02 localize to the liver after intravenous administration in mice; intravenous administration of LNPs containing OF-02 and encapsulating human EPO mRNA increases serum EPO levels in mice
Catalog No. 50-291-7954