Learn More
Cayman Chemical ANTIBODY OF-1 5mg
SDP
Supplier:  Cayman Chemical 171245 MG
Encompass_Preferred
A chemical probe for the bromodomains of the BRPF family of proteins; binds BRPF1B, BRPF2, and BRPF3 with Kd values of 0.1, 0.,5 and 2.4 µM, respectively, as determined by ITC
Catalog No. 50-296-5694