Learn More
Cayman Chemical ANTIBODY OF-Deg-LIn 5mg
SDP
Supplier:  Cayman Chemical 378535 MG
Encompass_Preferred
An ionizable cationic lipid; has been used in the generation of LNPs for the delivery of mRNA in vitro
Catalog No. 50-291-6827