Learn More
Vector Laboratories Protein ̶ Protein Conjugation A-9202-001
SDP
Supplier:  Vector Laboratories A9202001
Encompass
Protein–Protein Conjugation Kit. This kit is designed to easily and efficiently conjugate any two proteins. This kit is flexible so that researchers with little or no bioconjugation experience can make their own custom protein–protein conjugates to suit their research needs. It includes all of the necessary components, including S-HyNic and S-4FB, for the rapid and specific crosslinking of any two proteins with molecular weights greater than 25 kDa. This kit contains sufficient reagents for the preparation of two protein–protein conjugates, with up to 650 µg of each protein.
Catalog No. 50-204-5717