Learn More
Cayman Chemical ANTIBODY on-4059 25mg
SDP
Supplier:  Cayman Chemical 3295725 MG
Encompass_Preferred
A BTK inhibitor (IC50 = 2.2 nM); selective for BTK over LCK, LYN, and Fyn at 1 µM; inhibits B cell proliferation and activation in vitro and reduces tumor growth in a TMD8 mouse xenograft model
Catalog No. 50-294-8073