Learn More
Cayman Chemical ANTIBODY OrotIdIn 10mg
SDP
Supplier:  Cayman Chemical 3640110 MG
Encompass_Preferred
A pyrimidine nucleoside; urine levels are increased in rats following administration of allopurinol
Catalog No. 50-291-1742