Learn More
Cayman Chemical ANTIBODY OSI-027 5mg
SDP
Supplier:  Cayman Chemical 173795 MG
Encompass_Preferred
An inhibitor of the catalytic site of mTOR, inhibiting both mTORC1 and mTORC2 (IC50s = 22 and 65 nM, respectively); prohibits proliferation, induces autophagy, and potentiates apoptosis in BCR-ABL transformed cells and other cancer cells at 10 µM; effective in vivo
Catalog No. 50-298-0303