Learn More
Cayman Chemical ANTIBODY OTX015 10mg
SDP
Supplier:  Cayman Chemical 1594710 MG
Encompass_Preferred
An orally bioavailable, small molecule inhibitor of BRD2, BRD3, and BRD4 (EC50s = 10-19 nM) that displays antiproliferative effects at a sub-micromolar range in vitro in a wide range of solid tumor types and leukemias
Catalog No. 50-297-1386