Learn More
Cayman Chemical ANTIBODY PamapImod 5mg
SDP
Supplier:  Cayman Chemical 209715 MG
Encompass_Preferred
A potent, selective inhibitor of p38α MAPK (IC50 = 14 nM); blocks inflammatory signaling in cells, whole blood, and synovial explants; orally bioavailable, abrogating inflammation, bone, and pain in animal models
Catalog No. 50-298-1559