Learn More
Cayman Chemical ANTIBODY PD 117519 50mg
SDP
Supplier:  Cayman Chemical 2844450 MG
Encompass_Preferred
An adenosine receptor agonist; selectively binds to adenosine A2 receptors (IC50 = 30 µM) over A1 receptors (IC50 = 810 µM) in rat brain membranes; increases heart rate and decreases systolic blood pressure in normotensive dogs at 2 and 10 mg/kg
Catalog No. 50-294-0882