Learn More
Cayman Chemical ANTIBODY PD 168393 1mg
SDP
Supplier:  Cayman Chemical 210591 MG
Encompass_Preferred
A cell-permeable, irreversible inhibitor of EGFR kinase activity (IC50 = 0.70 nM); used in cells and in vivo to irreversibly inhibit EGFR signaling
Catalog No. 50-298-4194