Learn More
Cayman Chemical ANTIBODY PD 174265 500ug
SDP
Supplier:  Cayman Chemical 17649500 UG
Encompass_Preferred
A potent, cell-permeable inhibitor of the tyrosine kinase activity of the epidermal growth factor (EGF) receptor (IC50 = 0.45 nM); effective in cells, blocking tyrosine phosphorylation induced by either EGF or heregulin (IC50s = 39 and 220 nM, respectively)
Catalog No. 50-298-0231