Learn More
Cayman Chemical ANTIBODY PelItInIb 25mg
SDP
Supplier:  Cayman Chemical 1562725 MG
Encompass_Preferred
A cyanoquinoline that irreversibly inhibits the receptor tyrosine kinases EGFR (IC50 = 39 nM) and HER2 (IC50 = 1.2 μM); disrupts Akt and MAPK pathways, inducing apoptosis and suppressing the growth of tumor cell lines
Catalog No. 50-296-9191