Learn More
Cayman Chemical ANTIBODY PEO-IAA 250mg
SDP
Supplier:  Cayman Chemical 40665250 MG
Encompass_Preferred
An auxin antagonist; slows shoot growth and suppresses auxin-induced apical dominance in C. sativa seedlings at 10 UG/L in growth media
Catalog No. 50-294-2989