Learn More
Cayman Chemical ANTIBODY PF-299804 5mg
SDP
Supplier:  Cayman Chemical 90018795 MG
Encompass_Preferred
An irreversible pan-ErbB receptor tyrosine kinase inhibitor (IC50s = 6, 45.7, and 73.7 nM for ErbB1, ErbB2, and ErbB4, respectively); demonstrates antitumor activity in various tumor xenograft models expressing either wild-type ErbB or mutant ErbB family members that show resistance to first generation ErbB kinase inhibitors
Catalog No. 50-289-4333