Learn More
Cayman Chemical ANTIBODY PF-3450074 5mg
SDP
Supplier:  Cayman Chemical 361075 MG
Encompass_Preferred
An inhibitor of HIV-1 capsid protein; reduces viral infectivity in HeLa cells infected with wild-type HIV-1 but not P38A, E45A, Q63A/Q67A, or Q219A capsid protein mutant HIV-1, simian immunodeficiency virus, or murine leukemia virus at 10 µM; reduces capsid protein levels in HIV-1 viral cores
Catalog No. 50-291-3196