Learn More
Cayman Chemical ANTIBODY PF-3716556 25mg
SDP
Supplier:  Cayman Chemical 1771025 MG
Encompass_Preferred
A potent and selective H+/K+-ATPase inhibitor; pIC50s = 6.026, 6.038, and 6.009, respectively, for porcine membrane vesicles, and canine and human recombinant cells in ion-leaky (acidic) conditions; 3-fold greater inhibition than revaprazan in ion-tight conditions
Catalog No. 50-296-6351