Learn More
Cayman Chemical ANTIBODY PF-998425 10mg
SDP
Supplier:  Cayman Chemical 3144210 MG
Encompass_Preferred
A non-steroidal androgen receptor antagonist (IC50 = 43 nM in a cell-based assay); binds to the androgen receptor (IC50 = 37 nM in a radioligand binding assay); selective for the androgen receptor over the progesterone receptor (IC50 = >10,000 nM); topical administration increases hair growth in a mouse model of hair growth at 3% twice per day, as well as reduces EAr sebum in male Syrian golden hamsters (ED50 = 0.4% twice per day)
Catalog No. 50-299-1723