Learn More
Cayman Chemical ANTIBODY PhenytoIn 50g
SDP
Supplier:  Cayman Chemical 2403750 G
Encompass_Preferred
An anticonvulsant agent and active metabolite of fosphenytoin; inhibits neuronal voltage-gated sodium channels in a voltage-dependent manner; reduces the neuronal firing frequency and decreases the amplitude of EPSPs in electrically stimulated rat corticostriatal slices (EC50s = 42.8 and 33.5 µM, respectively); protects against MES-induced seizures in mice (ED50 = 10 mg/kg)
Catalog No. 50-293-3283