Learn More
Cayman Chemical ANTIBODY PhloxIn B 10g
SDP
Supplier:  Cayman Chemical 1867510 G
Encompass_Preferred
A red dye that can be used as an alternative to Gram staining; inhibits MRSA growth in vitro at 100 UG/ml; modulates channel activity of wild-type CFTR (Ki = 38 µM) and mutant delta F508 CFTR (Ki = 33 µM)
Catalog No. 50-297-9979