Learn More
Cayman Chemical ANTIBODY PHOP 500UG
SDP
Supplier:  Cayman Chemical 71655500 UG
Encompass_Preferred
A potent FAAH inhibitor, exhibiting Ki values of 0.094 and 0.2 nM for the human and rat enzymes, respectively
Catalog No. 50-289-6508