Learn More
Cayman Chemical ANTIBODY PIK-293 10mg
SDP
Supplier:  Cayman Chemical 2128310 MG
Encompass_Preferred
A selective inhibitor of the PI3K catalytic subunit p110δ (IC50s = 0.24 µM)
Catalog No. 50-298-1247