Learn More
Cayman Chemical ANTIBODY PIK-III 1mg
SDP
Supplier:  Cayman Chemical 170021 MG
Encompass_Preferred
A selective inhibitor of Vps34 (PI3K type 3; IC50 = 18 nM); acutely inhibits autophagy and de novo lipidation of LC3, leading to stabilization of autophagy substrates
Catalog No. 50-296-4989