Learn More
Cayman Chemical ANTIBODY PIroxIcam 10g
SDP
Supplier:  Cayman Chemical 1336810 G
Encompass_Preferred
A COX inhibitor and NSAID; inhibits TXB2 production from arachidonic acid in HEL human erythroleukemic cells (IC50 = 0.45 µM), as well as inhibits LPS-induced formation of PGF1α from arachidonic acid in Mono-Mac-6 cells (IC50 = 0.77 µM); reduces LPS-induced production of nitric oxide and IL-6 in cell-based assays (IC50s = 240 and ~470 µM, respectively); reduces carrageenan-induced paw edema in rats at 1, 2.5, and 5 mg/kg
Catalog No. 50-295-5380