Learn More
Cayman Chemical ANTIBODY PIrozadIl 250mg
SDP
Supplier:  Cayman Chemical 37292250 MG
Encompass_Preferred
A lipid-lowering agent; reduces serum and aortic cholesterol levels, serum LDL levels, and atherosclerotic lesion areas on the aorta and coronary lumen in a rabbit model of atherosclerosis induced by a high-cholesterol diet at 50 mg/kg per day
Catalog No. 50-291-8010