Learn More
Cayman Chemical ANTIBODY PK-THPP 10mg
SDP
Supplier:  Cayman Chemical 2947310 MG
Encompass_Preferred
A K2P9.1/TASK-3 inhibitor (IC50 = 0.035 µM); selective for K2P9.1/TASK-3 over K2P2.1/TREK1 and Kv1.5 (IC50s = ~5 and >10 µM, respectively), as well as a panel of more than 100 receptors, ion channels, and enzymes at 10 µM but does inhibit K2P3.1/TASK-1 (IC50 = 0.3 µM); increases the time spent awake and decreases the duration of REM and delta sleep in mice at 100 mg/kg
Catalog No. 50-299-1238