Learn More
Cayman Chemical ANTIBODY PKSI-527 25mg
SDP
Supplier:  Cayman Chemical 2397425 MG
Encompass_Preferred
An inhibitor of plasma kallikrein (Ki = 0.81 μM); reduces bradykinin generation induced by kaolin and λ-carrageenan ex vivo in human plasma; prolongs partial thromboplastin and euglobulin clot lysis times; reduces hyperplasia, pannus formation, and infiltration of inflammatory cells in the tarsal joint of mice with collagen-induced arthritis
Catalog No. 50-293-3202