Learn More
Cayman Chemical ANTIBODY PoloppIn 25mg
SDP
Supplier:  Cayman Chemical 3833025 MG
Encompass_Preferred
An inhibitor of the Plk1-Plk1-binding phosphopeptide protein-peptide interaction (IC50 = 26.9 µM); induces mitotic arrest in HeLa cells (EC50 = 29.9 µM); inhibits cell growth in murine pancreatic organoids expressing mutant K-Ras (GI50s = 22.92-28.39 µM)
Catalog No. 50-291-8116