Learn More
Cayman Chemical ANTIBODY PolygodIal 1mg
SDP
Supplier:  Cayman Chemical 149791 MG
Encompass_Preferred
A sesquiterpene dialdehyde noted for its broad antifungal properties; cytotoxic against bacteria, algae, and sea squirts; in mammals, produces a pungent flavor, activates TRPA1 (EC50 = 59 nM), and produces antinociception
Catalog No. 50-297-0644