Learn More
Cayman Chemical ANTIBODY PR-619 50mg
SDP
Supplier:  Cayman Chemical 1627650 MG
Encompass_Preferred
A general inhibitor of DUBs, significantly reducing the activity of a panel of DUBs with EC50 values ranging from 5-20 µM; cell-permeable and reversible in its action
Catalog No. 50-297-0031