Learn More
Cayman Chemical ANTIBODY Prasugrel 100mg
SDP
Supplier:  Cayman Chemical 14278100 MG
Encompass_Preferred
A prodrug form of R-99224; inhibits ex vivo washed platelet aggregation in rat platelet rich-plasma at 0.3 and 3 mg/kg; prevents thrombus formation (ED50 = 0.68 mg/kg) and increases tail bleeding time in rats
Catalog No. 50-295-5345