Learn More
Cayman Chemical ANTIBODY PrImIdon 1mg
SDP
Supplier:  Cayman Chemical 192771 MG
Encompass_Preferred
An analytical reference standard categorized as a barbiturate; partially substitutes for barbitone and reduces barbitone withdrawal-induced convulsions in rats in a model of barbitone dependence; intended for research and forensic applications
Catalog No. 50-298-1155