Learn More
Cayman Chemical ANTIBODY PRN-1371 10mg
SDP
Supplier:  Cayman Chemical 3378710 MG
Encompass_Preferred
An irreversible pan-FGFR inhibitor (IC50s = 0.7, 1.3, 4.4, and 19.3 nM for FGFR-1, -2, -3, and -4, respectively); selective for FGFR over VEGFR2 (IC50 = 705 nM) and a panel of 250 kinases (IC50s = >1 µM for all) but does inhibit CSF1R activity by >90% at 1 µM; inhibits proliferation in a panel of ten cancer cell lines containing various FGFR mutants (IC50s = 2-231 nM) and induces apoptosis in SNU-16 gastric and RT4 bladder cancer cells (EC50s = 15.9 and 11.8 nM, respectively); reduces tumor growth in an SNU-16 mouse xenograft model and a PDX mouse model of liver cancer at 15 mg/kg twice per day
Catalog No. 50-294-7031