Learn More
Cayman Chemical ANTIBODY PSB-KD107 10mg
SDP
Supplier:  Cayman Chemical 3139310 MG
Encompass_Preferred
A GPR18 agonist (EC50 = 0.56 µM in a β-arrestin recruitment assay using CHO cells expressing the human receptor); selective for GPR18 over GPR55, as well as cannabinoid 1 (CB1) and CB2 receptors, at 10 µM but also binds to the adenosine A2A receptor (Ki = 0.33 µM for the rat receptor)
Catalog No. 50-299-1875