Learn More
Cayman Chemical ANTIBODY PSB-SB1202 10mg
SDP
Supplier:  Cayman Chemical 1562110 MG
Encompass_Preferred
A CB1 and CB2 agonist with EC50 values of 56 and 14 nM, respectively; selective for the CB1 and CB2 receptors (Kis = 32 and 49 nM, respectively) over GPR55 (IC50 = 6.4 µM)
Catalog No. 50-297-2123