Learn More
Cayman Chemical ANTIBODY PsoralIdIn 10mg
SDP
Supplier:  Cayman Chemical 1511910 MG
Encompass_Preferred
A furanocoumarin isolated from the seeds of P. corylifolia that has been shown to induce cytotoxicity against various cancer cells through TRAIL-mediated events
Catalog No. 50-296-8676