Learn More
Cayman Chemical ANTIBODY PsoralIdIn 5mg
SDP
Supplier:  Cayman Chemical 151195 MG
Encompass_Preferred
A furanocoumarin isolated from the seeds of P. corylifolia that has been shown to induce cytotoxicity against various cancer cells through TRAIL-mediated events
Catalog No. 50-297-0787