Learn More
Cayman Chemical ANTIBODY PTIO 100mg
SDP
Supplier:  Cayman Chemical 14982100 MG
Encompass_Preferred
An oxidizing reagent that reacts with nitric oxide to form nitric dioxide and corresponding imino nitroxides
Catalog No. 50-297-2866