Learn More
Abcam Antibody Purification Kit (Protein A), 1 Tests
Supplier:  Abcam AB1027841TEST
Encompass_Preferred
Commercially available antibodies often contain substances (e.g. BSA, glycine, tris, azide) that interfere in labeling reactions with enzymes or fluorophores. ab102784 quickly removes these contaminants.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0859740