Learn More
Abcam Antibody Purification Kit (Gold/Europium labelling), 3 Tests
Supplier:  Abcam AB2049093TEST
Encompass_Preferred
Antibody Purification Kit - Nanoparticles (ab204909) removes common buffer constituents found in commercial antibodies.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-7638