Learn More
Cayman Chemical ANTIBODY Pyrazolam 10mg
SDP
Supplier:  Cayman Chemical 1490110 MG
Encompass_Preferred
An analytical reference standard categorized as a benzodiazepine; has been abused recreationally and has anxiolytic-like effects in mice; intended for research and forensic applications
Catalog No. 50-296-9405