Learn More
Cayman Chemical ANTIBODY PyrIdoxIn 50g
SDP
Supplier:  Cayman Chemical 2070650 G
Encompass_Preferred
A 4-methanol form of vitamin B6; has been used in cell culture media as a precursor to pyridoxal 5’-phosphate
Catalog No. 50-298-2031