Learn More
Cayman Chemical ANTIBODY QuazInon 25mg
SDP
Supplier:  Cayman Chemical 2915125 MG
Encompass_Preferred
A selective PDE3 inhibitor with positive inotropic and vasodilating properties; induces relaxation of precontracted isolated human cavernous smooth muscle (IC50 = 4.2 μM); increases myocardial contractile force in anesthetized open-chest dogs in a dose-dependent manner at 10-300 μg/kg, as well as decreases systolic and diastolic blood pressure; inhibits DNA synthesis induced by PDGF-BB in bovine coronary artery smooth muscle cells in a concentration-dependent manner
Catalog No. 50-294-1954