Learn More
Cayman Chemical ANTIBODY QX-77 25mg
SDP
Supplier:  Cayman Chemical 2990325 MG
Encompass_Preferred
An activator of chaperone-mediated autophagy; decreases paraquat- or oleic acid-induced MEF and Neuro2a cell death; rescues defective trafficking and lysosomal localization of the CMA receptor LAMP2A in Ctns-/- MEFs and CTNS knockout human proximal tubule cells at 20 µM
Catalog No. 50-299-0223