Learn More
Cayman Chemical ANTIBODY R-7050 5mg
SDP
Supplier:  Cayman Chemical 168705 MG
Encompass_Preferred
A cell-permeable antagonist of signaling through the TNF-α receptor complex (EC50 = 0.631 µM), blocking TNF-α-induced binding of TNF-α receptor I with TRADD and RIP1
Catalog No. 50-296-7470