Learn More
Cayman Chemical ANTIBODY RAF265 1mg
SDP
Supplier:  Cayman Chemical 169911 MG
Encompass_Preferred
A dual inhibitor of mutant B-RafV600E and VEGFR2 (EC50s = 0.14 and 0.19 µM, respectively, in cell-based assays) that also inhibits wild-type B-Raf, PDGFR, CSF1R, RET, c-KIT, SRC, and STE20 (IC50s = 20-100 nM)
Catalog No. 50-296-6924