Learn More
Cayman Chemical ANTIBODY ReserpIn 1g
SDP
Supplier:  Cayman Chemical 164741 G
Encompass_Preferred
An alkaloid that irreversibly inhibits both human VMAT1 and VMAT2 (Ki = 34 and 12 nM, respectively); used to experimentally deplete monoamines in animals; also inhibits P-glycoprotein (IC50 = 0.5 µM)
Catalog No. 50-297-0952