Learn More
Cayman Chemical ANTIBODY RetIculol 5mg
SDP
Supplier:  Cayman Chemical 231485 MG
Encompass_Preferred
A cAMP phosphodiesterase inhibitor (IC50 = 41 µM); inhibits DNA topoisomerase I at a concentration of 45 µM in B16F10 melanoma cells; has antifungal activity against T. mentagrophyes with a MIC 1.8 µM; cytotoxic against A427 human lung and B16F10 mouse melanoma cells at concentrations of 25 and 200 µM, respectively; inhibits lung metastasis in mice
Catalog No. 50-299-5571