Learn More
Cayman Chemical ANTIBODY RGB-286638 250mg
SDP
Supplier:  Cayman Chemical 29499250 MG
Encompass_Preferred
A multi-kinase inhibitor; inhibits a variety of kinases, including CDK2-7 and CDK9, FMS, JAK2, c-Src, GSK3β, TAK1, JNK1A1, JNK1A2, AMPK, and MEK1 (IC50s = 1-55 nM); inhibits proliferation of multiple myeloma cancer cell lines endogenously expressing mutant and wild-type p53 (EC50s = 20-70 nM), as well as patient-derived multiple myeloma cells at 50 and 100 nM; induces G1/S and G2/M cell cycle arrest and apoptosis of MM.1S human multiple myeloma cells at 50 nM; decreases tumor growth in an MM.1S mouse xenograft model at 30 and 40 mg/kg per day for 14 days
Catalog No. 50-299-0030