Learn More
Cayman Chemical ANTIBODY RGFP109 5mg
SDP
Supplier:  Cayman Chemical 215685 MG
Encompass_Preferred
An HDAC1 and HDAC3 inhibitor (IC50s = 60 and 50 nM; Kis = 32 and 5 nM, respectively); upregulates frataxin mRNA levels in cultured cells obtained from Friedreich’s ataxia patients and in a GAA repeat-based mouse model of Friedreich’s ataxia
Catalog No. 50-298-7450